missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorotriisopropylsilane, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 221260500

 View more versions of this product

Catalog No. AC221260500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chlorotriisopropylsilane
Colorless to Yellow
198.0°C
Authentic
Glass bottle
1.4520 to 1.4540
MFCD00009656
0.902
Solubility in water: reacts
CC(C)[Si](Cl)(C(C)C)C(C)C
192.80
192.81
Liquid
13154-24-0
0.9020g/mL
63°C
96% min. (GC)
C9H21ClSi
[(CH3)2CH]3SiCl
50 g
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
KQIADDMXRMTWHZ-UHFFFAOYSA-N
chlorotris(propan-2-yl)silane
139400
97%

Chemical Identifiers

13154-24-0
192.80
KQIADDMXRMTWHZ-UHFFFAOYSA-N
139400
C9H21ClSi
MFCD00009656
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
CC(C)[Si](Cl)(C(C)C)C(C)C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only