Learn More
Chloroxynil, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5043203
Description
Chloroxynil is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChloroxynil is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications
Chloroxynil | |
143°C to 144°C | |
MFCD00002177 | |
UN3439 | |
Slightly soluble in water. | |
OC1=C(Cl)C=C(C=C1Cl)C#N | |
188.01 | |
188.01 |
1891-95-8 | |
C7H3Cl2NO | |
1 g | |
chloroxynil, benzonitrile, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzonitrile, chloroxynil iso, caswell no. 309a, unii-gi6x21wsvn, gi6x21wsvn, epa pesticide chemical code 309500, 4-cyano-2,6-dichlorophenol, 3,5-dichloro-4-hydroxy benzonitrile | |
YRSSHOVRSMQULE-UHFFFAOYSA-N | |
3,5-dichloro-4-hydroxybenzonitrile | |
74685 | |
≥97% |
Chemical Identifiers
1891-95-8 | |
188.01 | |
YRSSHOVRSMQULE-UHFFFAOYSA-N | |
74685 | |
OC1=C(Cl)C=C(C=C1Cl)C#N |
C7H3Cl2NO | |
MFCD00002177 | |
chloroxynil, benzonitrile, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzonitrile, chloroxynil iso, caswell no. 309a, unii-gi6x21wsvn, gi6x21wsvn, epa pesticide chemical code 309500, 4-cyano-2,6-dichlorophenol, 3,5-dichloro-4-hydroxy benzonitrile | |
3,5-dichloro-4-hydroxybenzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 217-572-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only