missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroxynil, 97+%, Thermo Scientific Chemicals

$268.30 - $1098.44

Chemical Identifiers

CAS 1891-95-8
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.01
MDL Number MFCD00002177
InChI Key YRSSHOVRSMQULE-UHFFFAOYSA-N
Synonym chloroxynil, benzonitrile, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzonitrile, chloroxynil iso, caswell no. 309a, unii-gi6x21wsvn, gi6x21wsvn, epa pesticide chemical code 309500, 4-cyano-2,6-dichlorophenol, 3,5-dichloro-4-hydroxy benzonitrile
PubChem CID 74685
IUPAC Name 3,5-dichloro-4-hydroxybenzonitrile
SMILES OC1=C(Cl)C=C(C=C1Cl)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5043203
View Documents
Thermo Scientific Chemicals
H5043203
1 g
Each for $268.30
 
AAH5043206
View Documents
Thermo Scientific Chemicals
H5043206
5 g
Each for $1,098.44
 
Description

Description

Chloroxynil is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroxynil is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications

Chemical Identifiers

1891-95-8
188.01
YRSSHOVRSMQULE-UHFFFAOYSA-N
74685
OC1=C(Cl)C=C(C=C1Cl)C#N
C7H3Cl2NO
MFCD00002177
chloroxynil, benzonitrile, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzonitrile, chloroxynil iso, caswell no. 309a, unii-gi6x21wsvn, gi6x21wsvn, epa pesticide chemical code 309500, 4-cyano-2,6-dichlorophenol, 3,5-dichloro-4-hydroxy benzonitrile
3,5-dichloro-4-hydroxybenzonitrile

Specifications

1891-95-8
C7H3Cl2NO
1 g
chloroxynil, benzonitrile, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzonitrile, chloroxynil iso, caswell no. 309a, unii-gi6x21wsvn, gi6x21wsvn, epa pesticide chemical code 309500, 4-cyano-2,6-dichlorophenol, 3,5-dichloro-4-hydroxy benzonitrile
YRSSHOVRSMQULE-UHFFFAOYSA-N
3,5-dichloro-4-hydroxybenzonitrile
74685
≥97%
143°C to 144°C
MFCD00002177
UN3439
Slightly soluble in water.
OC1=C(Cl)C=C(C=C1Cl)C#N
188.01
188.01
Chloroxynil
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 217-572-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only