Learn More
Chromium(III) nitrate nonahydrate, 99%, Thermo Scientific Chemicals
$92.15 - $454.46
Chemical Identifiers
CAS | 7789-02-8 |
---|---|
MDL Number | MFCD00149659 |
InChI Key | PHFQLYPOURZARY-UHFFFAOYSA-N |
Synonym | chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate |
PubChem CID | 9887206 |
IUPAC Name | chromium(3+);trinitrate;nonahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC219201000
|
Thermo Scientific Chemicals
219201000 |
100 g | Plastic bottle |
Each for $92.15
|
|
||||
AC219205000
|
Thermo Scientific Chemicals
219205000 |
500 g | Plastic bottle |
Each for $267.65
|
|
||||
AC219200010
|
Thermo Scientific Chemicals
219200010 |
1 kg | Plastic bottle |
Each for $454.46
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-02-8 | |
PHFQLYPOURZARY-UHFFFAOYSA-N | |
9887206 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3] |
MFCD00149659 | |
chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate | |
chromium(3+);trinitrate;nonahydrate |
Specifications
60.0°C | |
Black to Purple | |
Crystalline Chunks | |
98.5% min. (Iodometry) | |
Cr(NO3)3·9H2O | |
14,2226 | |
Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3] | |
400.14 | |
400.14 | |
Plastic bottle |
7789-02-8 | |
2 to 3 (5% aq. soln.) | |
100 g | |
CrN3O9·9H2O | |
MFCD00149659 | |
chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate | |
PHFQLYPOURZARY-UHFFFAOYSA-N | |
chromium(3+);trinitrate;nonahydrate | |
9887206 | |
99% | |
Chromium(III) nitrate nonahydrate, 99% |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
May intensify fire; oxidizer.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
Call a POISON CE
GHS Signal Word: Warning
RTECSNumber : GB6300000
RUO – Research Use Only