Learn More
cis-2,6-Dimethylmorpholine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals AC371845000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
cis-2, 6-Dimethylmorpholine | |
0.9300g/mL | |
41°C | |
2% max. Trans-2,6-dimethylmorpholine (GC) | |
C6H13NO | |
MFCD00078428 | |
0.93 | |
Solubility in water: miscible | |
CC1CNCC(O1)C | |
115.18 | |
5.3 mPa.s (20°C) | |
97% |
6485-55-8 | |
140°C to 142°C | |
Authentic | |
Glass bottle | |
1.4450 to 1.447 | |
500g | |
cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624 | |
HNVIQLPOGUDBSU-OLQVQODUSA-N | |
(2S,6R)-2,6-dimethylmorpholine | |
641500 | |
115.18 |
Chemical Identifiers
6485-55-8 | |
115.18 | |
HNVIQLPOGUDBSU-OLQVQODUSA-N | |
641500 | |
CC1CNCC(O1)C |
C6H13NO | |
MFCD00078428 | |
cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624 | |
(2S,6R)-2,6-dimethylmorpholine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa
GHS Signal Word: Danger
EINECSNumber : 229-353-
RUO â Research Use Only