missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-2,6-Dimethylmorpholine, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 6485-55-8 |
---|---|
Molecular Formula | C6H13NO |
Molecular Weight (g/mol) | 115.18 |
MDL Number | MFCD00078428 |
InChI Key | HNVIQLPOGUDBSU-OLQVQODUSA-N |
Synonym | cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624 |
PubChem CID | 641500 |
IUPAC Name | (2S,6R)-2,6-dimethylmorpholine |
SMILES | CC1CNCC(O1)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC371840250
|
Thermo Scientific Chemicals
371840250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC371841000
|
Thermo Scientific Chemicals
371841000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC371845000
|
Thermo Scientific Chemicals
AC371845000 |
500g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
6485-55-8 | |
115.18 | |
HNVIQLPOGUDBSU-OLQVQODUSA-N | |
641500 | |
CC1CNCC(O1)C |
C6H13NO | |
MFCD00078428 | |
cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624 | |
(2S,6R)-2,6-dimethylmorpholine |
Specifications
6485-55-8 | |
140°C to 142°C | |
Authentic | |
Glass bottle | |
1.4450 to 1.447 | |
25g | |
cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624 | |
HNVIQLPOGUDBSU-OLQVQODUSA-N | |
(2S,6R)-2,6-dimethylmorpholine | |
641500 | |
115.18 | |
cis-2, 6-Dimethylmorpholine |
0.9300g/mL | |
41°C | |
2% max. Trans-2,6-dimethylmorpholine (GC) | |
C6H13NO | |
MFCD00078428 | |
0.93 | |
Solubility in water: miscible | |
CC1CNCC(O1)C | |
115.18 | |
5.3 mPa.s (20°C) | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa
GHS Signal Word: Danger
EINECSNumber : 229-353-
RUO â Research Use Only