missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-2-Butene-1,4-diol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 107720025

 View more versions of this product

Catalog No. AC107720025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

cis-2-Butene-1, 4-diol
6117-80-2
Colorless
235.0°C
Authentic
Glass bottle
1.4770 to 1.4800
MFCD00002924,MFCD00063207
1.07
Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone,slightly soluble in benzene
OC\C=C/CO
88.11
88.11
Liquid
97% cis
7.0°C
1.0700g/mL
128°C
96% min. (cis-isomer) (GC)
C4H8O2
HOCH2CH=CHCH2OH
2.5 L
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv
ORTVZLZNOYNASJ-UPHRSURJSA-N
(Z)-but-2-ene-1,4-diol
643790
97%

Chemical Identifiers

6117-80-2
88.11
ORTVZLZNOYNASJ-UPHRSURJSA-N
643790
C4H8O2
MFCD00002924,MFCD00063207
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv
OC\C=C/CO
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove

GHS Signal Word: Warning

EINECSNumber : 228-085-1

Recommended Storage : below 6°C, crystals may form, agitation necessary to disperse

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only