Learn More
cis-2-Butene-1,4-diol, 97%, Thermo Scientific Chemicals
$70.35 - $873.17
Chemical Identifiers
CAS | 6117-80-2 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.11 |
MDL Number | MFCD00002924,MFCD00063207 |
InChI Key | ORTVZLZNOYNASJ-UPHRSURJSA-N |
Synonym | cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv |
PubChem CID | 643790 |
SMILES | OC\C=C/CO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC107721000
|
Thermo Scientific Chemicals
107721000 |
100 mL | Glass bottle |
Each for $70.35
|
|
||||
AC107725000
|
Thermo Scientific Chemicals
107725000 |
500 mL | Glass bottle |
Each for $212.63
|
|
||||
AC107720025
|
Thermo Scientific Chemicals
107720025 |
2.5 L | Glass bottle |
Each for $873.17
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6117-80-2 | |
88.11 | |
ORTVZLZNOYNASJ-UPHRSURJSA-N | |
643790 |
C4H8O2 | |
MFCD00002924,MFCD00063207 | |
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv | |
OC\C=C/CO |
Specifications
6117-80-2 | |
Colorless | |
235.0°C | |
Authentic | |
Glass bottle | |
1.4770 to 1.4800 | |
MFCD00002924,MFCD00063207 | |
1.07 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone,slightly soluble in benzene | |
OC\C=C/CO | |
88.11 | |
88.11 | |
Liquid |
7.0°C | |
1.0700g/mL | |
128°C | |
96% min. (cis-isomer) (GC) | |
C4H8O2 | |
HOCH2CH=CHCH2OH | |
100 mL | |
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv | |
ORTVZLZNOYNASJ-UPHRSURJSA-N | |
(Z)-but-2-ene-1,4-diol | |
643790 | |
97% | |
cis-2-Butene-1, 4-diol, 97% cis |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
GHS Signal Word: Warning
EINECSNumber : 228-085-1
Recommended Storage : below 6°C, crystals may form, agitation necessary to disperse
RUO – Research Use Only