Learn More
cis-2-Butene-1,4-diol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 107725000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
cis-2-Butene-1, 4-diol | |
6117-80-2 | |
Colorless | |
235.0°C | |
Authentic | |
Glass bottle | |
1.4770 to 1.4800 | |
MFCD00002924,MFCD00063207 | |
1.07 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone,slightly soluble in benzene | |
OC\C=C/CO | |
88.11 | |
88.11 | |
Liquid |
97% cis | |
7.0°C | |
1.0700g/mL | |
128°C | |
96% min. (cis-isomer) (GC) | |
C4H8O2 | |
HOCH2CH=CHCH2OH | |
500 mL | |
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv | |
ORTVZLZNOYNASJ-UPHRSURJSA-N | |
(2Z)-but-2-ene-1,4-diol | |
643790 | |
97% |
Chemical Identifiers
6117-80-2 | |
88.11 | |
ORTVZLZNOYNASJ-UPHRSURJSA-N | |
643790 |
C4H8O2 | |
MFCD00002924,MFCD00063207 | |
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv | |
OC\C=C/CO |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
GHS Signal Word: Warning
EINECSNumber : 228-085-1
Recommended Storage : below 6°C, crystals may form, agitation necessary to disperse
RUO – Research Use Only