missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-4-Decenal, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0638206

 View more versions of this product

Catalog No. AAL0638206



Description

Description

cis-4-Decenal is suitable for use as standard compound to investigate the release of odorants in hydrocolloid model systems containing original or regio-selectively carboxylated cellulose at different pH values using static headspace gas chromatographic (SHS-GC) analysis. It may be used as standard for the standardization of 9-decenal by gas-chromatography. It may be used in the preparation of cis-4-decenol. It has been identified as one of the volatile biological marker for grey mold (Botrytis cinerea) infections in strawberry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-Decenal is suitable for use as standard compound to investigate the release of odorants in hydrocolloid model systems containing original or regio-selectively carboxylated cellulose at different pH values using static headspace gas chromatographic (SHS-GC) analysis. It may be used as standard for the standardization of 9-decenal by gas-chromatography. It may be used in the preparation of cis-4-decenol. It has been identified as one of the volatile biological marker for grey mold (Botrytis cinerea) infections in strawberry.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

cis-4-Decenal
0.857
81°C (177°F)
1.444
5 g
trans-4-decen-1-al, trans-4-decenal, e-4-decenal, e-dec-4-enal, 4-decenal, e, 4-decenal, unii-kz74b5801j, 4-decene-1-al, dec-4-enal, fema no. 3264
CWRKZMLUDFBPAO-VOTSOKGWSA-N
(E)-dec-4-enal
5702654
95%
21662-09-9
78°C to 80°C (10 mmHg)
C10H18O
MFCD00007024
2323646
Not miscible in water.
CCCCCC=CCCC=O
154.253
154.25

Chemical Identifiers

21662-09-9
154.253
CWRKZMLUDFBPAO-VOTSOKGWSA-N
5702654
CCCCCC=CCCC=O
C10H18O
MFCD00007024
trans-4-decen-1-al, trans-4-decenal, e-4-decenal, e-dec-4-enal, 4-decenal, e, 4-decenal, unii-kz74b5801j, 4-decene-1-al, dec-4-enal, fema no. 3264
(E)-dec-4-enal
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H227-H315-H319

EINECSNumber : 244-514-5

RTECSNumber : HE2071400

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only