Learn More
cis-4-Decenal, 95%, Thermo Scientific Chemicals
$52.10 - $122.18
Chemical Identifiers
CAS | 21662-09-9 |
---|---|
Molecular Formula | C10H18O |
Molecular Weight (g/mol) | 154.253 |
MDL Number | MFCD00007024 |
InChI Key | CWRKZMLUDFBPAO-VOTSOKGWSA-N |
Synonym | trans-4-decen-1-al, trans-4-decenal, e-4-decenal, e-dec-4-enal, 4-decenal, e, 4-decenal, unii-kz74b5801j, 4-decene-1-al, dec-4-enal, fema no. 3264 |
PubChem CID | 5702654 |
IUPAC Name | (E)-dec-4-enal |
SMILES | CCCCCC=CCCC=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0638203
|
Thermo Scientific Chemicals
L0638203 |
1 g |
Each for $52.10
|
|
|||||
AAL0638206
|
Thermo Scientific Chemicals
L0638206 |
5 g |
Each for $122.18
|
|
|||||
Description
cis-4-Decenal is suitable for use as standard compound to investigate the release of odorants in hydrocolloid model systems containing original or regio-selectively carboxylated cellulose at different pH values using static headspace gas chromatographic (SHS-GC) analysis. It may be used as standard for the standardization of 9-decenal by gas-chromatography. It may be used in the preparation of cis-4-decenol. It has been identified as one of the volatile biological marker for grey mold (Botrytis cinerea) infections in strawberry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationscis-4-Decenal is suitable for use as standard compound to investigate the release of odorants in hydrocolloid model systems containing original or regio-selectively carboxylated cellulose at different pH values using static headspace gas chromatographic (SHS-GC) analysis. It may be used as standard for the standardization of 9-decenal by gas-chromatography. It may be used in the preparation of cis-4-decenol. It has been identified as one of the volatile biological marker for grey mold (Botrytis cinerea) infections in strawberry.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
21662-09-9 | |
154.253 | |
CWRKZMLUDFBPAO-VOTSOKGWSA-N | |
5702654 | |
CCCCCC=CCCC=O |
C10H18O | |
MFCD00007024 | |
trans-4-decen-1-al, trans-4-decenal, e-4-decenal, e-dec-4-enal, 4-decenal, e, 4-decenal, unii-kz74b5801j, 4-decene-1-al, dec-4-enal, fema no. 3264 | |
(E)-dec-4-enal |
Specifications
21662-09-9 | |
78°C to 80°C (10 mmHg) | |
C10H18O | |
MFCD00007024 | |
2323646 | |
Not miscible in water. | |
CCCCCC=CCCC=O | |
154.253 | |
154.25 | |
cis-4-Decenal |
0.857 | |
81°C (177°F) | |
1.444 | |
1 g | |
trans-4-decen-1-al, trans-4-decenal, e-4-decenal, e-dec-4-enal, 4-decenal, e, 4-decenal, unii-kz74b5801j, 4-decene-1-al, dec-4-enal, fema no. 3264 | |
CWRKZMLUDFBPAO-VOTSOKGWSA-N | |
(E)-dec-4-enal | |
5702654 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
EINECSNumber : 244-514-5
RTECSNumber : HE2071400
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only