Learn More
cis-4-Octene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04315618
Description
cis-4-octene is used as a chain transfer agent in polymerization reactions, and as a depolymerizing agent in epoxy-functionalized oligomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
cis-4-Octene | |
C8H16 | |
50 g | |
cis-4-octene, z-oct-4-ene, z-4-octene, 4-octene, z, unii-19430whf5e, 4z-4-octene, 4z-oct-4-ene, 4-octene, 4z, cis-4-octene 1g, unii-68k818w2zc component | |
IRUCBBFNLDIMIK-FPLPWBNLSA-N | |
(Z)-oct-4-ene | |
5364446 | |
97% |
7642-15-1 | |
MFCD00067484 | |
UN3295 | |
Immiscible with water. | |
CCC\C=C/CCC | |
112.22 | |
112.22 | |
Liquid |
Chemical Identifiers
7642-15-1 | |
112.22 | |
IRUCBBFNLDIMIK-FPLPWBNLSA-N | |
5364446 |
C8H16 | |
MFCD00067484 | |
cis-4-octene, z-oct-4-ene, z-4-octene, 4-octene, z, unii-19430whf5e, 4z-4-octene, 4z-oct-4-ene, 4-octene, 4z, cis-4-octene 1g, unii-68k818w2zc component | |
CCC\C=C/CCC |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 231-578-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only