Learn More
cis-4-Octene, 97%, Thermo Scientific Chemicals
$76.16 - $1033.77
Chemical Identifiers
CAS | 7642-15-1 |
---|---|
Molecular Formula | C8H16 |
Molecular Weight (g/mol) | 112.22 |
MDL Number | MFCD00067484 |
InChI Key | IRUCBBFNLDIMIK-FPLPWBNLSA-N |
Synonym | cis-4-octene, z-oct-4-ene, z-4-octene, 4-octene, z, unii-19430whf5e, 4z-4-octene, 4z-oct-4-ene, 4-octene, 4z, cis-4-octene 1g, unii-68k818w2zc component |
PubChem CID | 5364446 |
SMILES | CCC\C=C/CCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4315604
|
Thermo Scientific Chemicals
04315604 |
2 g |
Each for $76.16
|
|
|||||
AA4315609
|
Thermo Scientific Chemicals
04315609 |
10 g |
Each for $255.81
|
|
|||||
AA4315618
|
Thermo Scientific Chemicals
04315618 |
50 g |
Each for $1,033.77
|
|
|||||
Description
cis-4-octene is used as a chain transfer agent in polymerization reactions, and as a depolymerizing agent in epoxy-functionalized oligomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7642-15-1 | |
112.22 | |
IRUCBBFNLDIMIK-FPLPWBNLSA-N | |
5364446 |
C8H16 | |
MFCD00067484 | |
cis-4-octene, z-oct-4-ene, z-4-octene, 4-octene, z, unii-19430whf5e, 4z-4-octene, 4z-oct-4-ene, 4-octene, 4z, cis-4-octene 1g, unii-68k818w2zc component | |
CCC\C=C/CCC |
Specifications
7642-15-1 | |
MFCD00067484 | |
UN3295 | |
Immiscible with water. | |
CCC\C=C/CCC | |
112.22 | |
112.22 | |
Liquid |
C8H16 | |
2 g | |
cis-4-octene, z-oct-4-ene, z-4-octene, 4-octene, z, unii-19430whf5e, 4z-4-octene, 4z-oct-4-ene, 4-octene, 4z, cis-4-octene 1g, unii-68k818w2zc component | |
IRUCBBFNLDIMIK-FPLPWBNLSA-N | |
(Z)-oct-4-ene | |
5364446 | |
97% | |
cis-4-Octene |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 231-578-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only