missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-Cyclooctene, 95%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 154862500

 View more versions of this product

Catalog No. AC154862500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

cis-Cyclooctene
-16.0°C
0.8400g/mL
25°C
94% min. (GC)
C8H14
C8H14
250 mL
cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene
URYYVOIYTNXXBN-UPHRSURJSA-N
cyclooctene
100 to 200ppm irganox 1076 FD
1.89 mPa.s (20°C)
95%
931-87-3
Colorless to Brown
145.0°C to 146.0°C
Authentic
Glass bottle
1.4680 to 1.4710
MFCD00001753
0.84
Solubility in water: <0.1g/L (20°C)
C1CCC\C=C/CC1
110.20
638079
110.2
Liquid

Chemical Identifiers

931-87-3
110.20
URYYVOIYTNXXBN-UPHRSURJSA-N
638079
C8H14
MFCD00001753
cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene
C1CCC\C=C/CC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
Store in a well

GHS Signal Word: Danger

EINECSNumber : 213-243-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only