missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-Cyclooctene, 95%, stabilized, Thermo Scientific Chemicals

$125.39 - $365.71

Chemical Identifiers

CAS 931-87-3
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00001753
InChI Key URYYVOIYTNXXBN-UPHRSURJSA-N
Synonym cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene
PubChem CID 638079
SMILES C1CCC\C=C/CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC154862500
SDS View Documents
Thermo Scientific Chemicals
154862500
250 mL Glass bottle
Each for $125.39
 
AC154860010
SDS View Documents
Thermo Scientific Chemicals
154860010
1 L Glass bottle
Each for $365.71
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

931-87-3
110.20
URYYVOIYTNXXBN-UPHRSURJSA-N
638079
C8H14
MFCD00001753
cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene
C1CCC\C=C/CC1

Specifications

931-87-3
Colorless to Brown
145.0°C to 146.0°C
Authentic
Glass bottle
1.4680 to 1.4710
MFCD00001753
0.84
Solubility in water: <0.1g/L (20°C)
C1CCC\C=C/CC1
110.20
638079
110.2
Liquid
-16.0°C
0.8400g/mL
25°C
94% min. (GC)
C8H14
C8H14
250 mL
cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene
URYYVOIYTNXXBN-UPHRSURJSA-N
cyclooctene
100 to 200ppm irganox 1076 FD
1.89 mPa.s (20°C)
95%
cis-Cyclooctene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
Store in a well

GHS Signal Word: Danger

EINECSNumber : 213-243-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only