missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 110440050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Citral | |
-10°C | |
0.8890g/mL | |
101°C | |
94% min. (GC) | |
C10H16O | |
(CH3)2C=CHCH2CH2C(CH3)=CHCHO | |
5 mL | |
15, 2321 | |
Solubility in water: practically insoluble. Other solubilities: soluble in 5 volumes of 60% alcohol,soluble in al,benzyl benzoate,diethyl phthalate,glycerol,prop,mineral oil,fixed oils | |
CC(C)=CCC\C(C)=C\C=O | |
152.24 | |
CHEBI:16980 | |
95% |
5392-40-5 | |
Yellow | |
229°C | |
Authentic | |
Glass bottle | |
1.4860 to 1.49 | |
MFCD00006997 | |
0.889 | |
citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb | |
WTEVQBCEXWBHNA-JXMROGBWSA-N | |
(2E)-3,7-dimethylocta-2,6-dienal | |
638011 | |
152.24 | |
Liquid |
Chemical Identifiers
5392-40-5 | |
152.24 | |
WTEVQBCEXWBHNA-JXMROGBWSA-N | |
638011 | |
(2E)-3,7-dimethylocta-2,6-dienal |
C10H16O | |
MFCD00006997 | |
citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb | |
CHEBI:16980 | |
CC(C)=CCC\C(C)=C\C=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 226-394-6
RTECSNumber : RG5075000
TSCA : TSCA
RUO – Research Use Only