missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals

$48.08 - $257.98

Chemical Identifiers

CAS 5392-40-5
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00006997
InChI Key WTEVQBCEXWBHNA-JXMROGBWSA-N
Synonym citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
PubChem CID 638011
ChEBI CHEBI:16980
IUPAC Name (2E)-3,7-dimethylocta-2,6-dienal
SMILES CC(C)=CCC\C(C)=C\C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC110440050
SDS View Documents
Thermo Scientific Chemicals
110440050
5 mL Glass bottle
Each for $48.08
 
AC110441000
SDS View Documents
Thermo Scientific Chemicals
110441000
100 mL Glass bottle
Each for $73.49
 
AC110445000
SDS View Documents
Thermo Scientific Chemicals
110445000
500 mL Glass bottle
Each for $257.98
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5392-40-5
152.24
WTEVQBCEXWBHNA-JXMROGBWSA-N
638011
(2E)-3,7-dimethylocta-2,6-dienal
C10H16O
MFCD00006997
citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
CHEBI:16980
CC(C)=CCC\C(C)=C\C=O

Specifications

5392-40-5
Yellow
229°C
Authentic
Glass bottle
1.4860 to 1.49
MFCD00006997
0.889
citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
WTEVQBCEXWBHNA-JXMROGBWSA-N
(2E)-3,7-dimethylocta-2,6-dienal
638011
152.24
Liquid
-10°C
0.8890g/mL
101°C
94% min. (GC)
C10H16O
(CH3)2C=CHCH2CH2C(CH3)=CHCHO
5 mL
15, 2321
Solubility in water: practically insoluble. Other solubilities: soluble in 5 volumes of 60% alcohol,soluble in al,benzyl benzoate,diethyl phthalate,glycerol,prop,mineral oil,fixed oils
CC(C)=CCC\C(C)=C\C=O
152.24
CHEBI:16980
95%
Citral
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 226-394-6

RTECSNumber : RG5075000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only