missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 110441000

 View more versions of this product

Catalog No. AC110441000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Citral
-10°C
0.8890g/mL
101°C
94% min. (GC)
C10H16O
(CH3)2C=CHCH2CH2C(CH3)=CHCHO
100 mL
15, 2321
Solubility in water: practically insoluble. Other solubilities: soluble in 5 volumes of 60% alcohol,soluble in al,benzyl benzoate,diethyl phthalate,glycerol,prop,mineral oil,fixed oils
CC(C)=CCC\C(C)=C\C=O
152.24
CHEBI:16980
95%
5392-40-5
Yellow
229°C
Authentic
Glass bottle
1.4860 to 1.49
MFCD00006997
0.889
citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
WTEVQBCEXWBHNA-JXMROGBWSA-N
(2E)-3,7-dimethylocta-2,6-dienal
638011
152.24
Liquid

Chemical Identifiers

5392-40-5
152.24
WTEVQBCEXWBHNA-JXMROGBWSA-N
638011
(2E)-3,7-dimethylocta-2,6-dienal
C10H16O
MFCD00006997
citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
CHEBI:16980
CC(C)=CCC\C(C)=C\C=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 226-394-6

RTECSNumber : RG5075000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only