missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citric Acid Monohydrate (Granular/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical FLA104250LB

 View more versions of this product

Catalog No. A104-250LB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Citric Acid Monohydrate
135°C to 152°C
175°C
99.0 to 102.0 %
(HO2CCH2)2C(OH)(CO2H)·H2O
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
210.138
CHEBI:31404
99.0 to 102.0%
1.54g/mL
174°C
Fiber Drum
2.2
5949-29-1
White
C6H10O8
MFCD00149972
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
22230
210.14
Certified ACS
250 lb.
0.02% max.
Solid

Chemical Identifiers

5949-29-1
210.138
YASYEJJMZJALEJ-UHFFFAOYSA-N
22230
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C6H10O8
MFCD00149972
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
CHEBI:31404
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Risk of serious damage to eyes. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:1
Instability:0

SDS Documents

Documents

Accessories
Promotions

Promotions