missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citric Acid Monohydrate (Granular/Certified ACS), Fisher Chemical™

$280.33 - $16762.33

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
MDL Number MFCD00149972
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A104-500 Edge
SDS View Documents Promotion Details
Fisher Chemical
FLA104500
500 g Poly Bottle
Each for $280.33
Case of 6 Each for $1,681.98
Edge
 
A104-250LB
SDS Accessories View Documents
Thermo Fisher Scientific
FLA104250LB
250 lb. Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A104-3
SDS View Documents Promotion Details
Thermo Fisher Scientific
FLA1043
3 kg Poly Bottle
Each for $841.08
Case of 4 Each for $3,364.32
 
A104-10
SDS View Documents
Thermo Fisher Scientific
FLA10410
10 kg Poly Pail
Each for $2,044.75
 
Description

Description

Specifications

Chemical Identifiers

5949-29-1
210.138
YASYEJJMZJALEJ-UHFFFAOYSA-N
22230
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C6H10O8
MFCD00149972
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
CHEBI:31404
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

Specifications

2.2
5949-29-1
White
C6H10O8
MFCD00149972
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
22230
210.14
Certified ACS
500 g
0.02% max.
Solid
135°C to 152°C
175°C
99.0 to 102.0 %
(HO2CCH2)2C(OH)(CO2H)·H2O
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
210.138
CHEBI:31404
99.0 to 102.0%
1.54g/mL
174°C
Poly Bottle
Citric Acid Monohydrate
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Risk of serious damage to eyes. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:1
Instability:0

SDS
missing translation for 'documents'

missing translation for 'documents'