missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 327160010

 View more versions of this product

Catalog No. AC327160010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Citric acid, trisodium salt hydrate
114456-61-0
>300.0°C
7 to 9 (5% aq. soln.)
Authentic
Glass Bottle
MFCD00150032
14, 8602
Solubility in water: soluble.
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
258.07
258.08
Molecular Biology
For molecular biology
0.02 max. (0.1 M in water) at 260nm
White
12% to 13%
0.998
C6H5Na3O7
1 kg
trisodium citrate hydrate, sodium citrate tribasic hydrate, citric acid trisodium salt hydrate, sodium citrate, tribasic, sodium citrate monohydrate, c6h5o7.3na.h2o, sodium citrate tribasic hydrate, reagentplus r, trisodium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
trisodium 2-hydroxypropane-1,2,3-tricarboxylate
21868242
0.998
Crystalline Powder or Crystals

Chemical Identifiers

114456-61-0
258.07
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
21868242
C6H5Na3O7
MFCD00150032
trisodium citrate hydrate, sodium citrate tribasic hydrate, citric acid trisodium salt hydrate, sodium citrate, tribasic, sodium citrate monohydrate, c6h5o7.3na.h2o, sodium citrate tribasic hydrate, reagentplus r, trisodium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only