missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and, Thermo Scientific Chemicals
$99.15 - $110.18
Chemical Identifiers
CAS | 114456-61-0 |
---|---|
Molecular Formula | C6H5Na3O7 |
Molecular Weight (g/mol) | 258.07 |
MDL Number | MFCD00150032 |
InChI Key | HRXKRNGNAMMEHJ-UHFFFAOYSA-K |
Synonym | trisodium citrate hydrate, sodium citrate tribasic hydrate, citric acid trisodium salt hydrate, sodium citrate, tribasic, sodium citrate monohydrate, c6h5o7.3na.h2o, sodium citrate tribasic hydrate, reagentplus r, trisodium 2-hydroxypropane-1,2,3-tricarboxylate hydrate |
PubChem CID | 21868242 |
SMILES | [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC327162500
|
Thermo Scientific Chemicals
327162500 |
250 g | Plastic bottle |
N/A
|
|
|||||
AC327160010
|
Thermo Scientific Chemicals
327160010 |
1 kg | Glass Bottle |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
114456-61-0 | |
258.07 | |
HRXKRNGNAMMEHJ-UHFFFAOYSA-K | |
21868242 |
C6H5Na3O7 | |
MFCD00150032 | |
trisodium citrate hydrate, sodium citrate tribasic hydrate, citric acid trisodium salt hydrate, sodium citrate, tribasic, sodium citrate monohydrate, c6h5o7.3na.h2o, sodium citrate tribasic hydrate, reagentplus r, trisodium 2-hydroxypropane-1,2,3-tricarboxylate hydrate | |
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
Specifications
114456-61-0 | |
>300.0°C | |
7 to 9 (5% aq. soln.) | |
Authentic | |
Plastic bottle | |
MFCD00150032 | |
14, 8602 | |
Solubility in water: soluble. | |
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O | |
258.07 | |
258.08 | |
Molecular Biology | |
Citric acid, trisodium salt hydrate, For molecular biology |
0.02 max. (0.1 M in water) at 260nm | |
White | |
12% to 13% | |
0.998 | |
C6H5Na3O7 | |
250 g | |
trisodium citrate hydrate, sodium citrate tribasic hydrate, citric acid trisodium salt hydrate, sodium citrate, tribasic, sodium citrate monohydrate, c6h5o7.3na.h2o, sodium citrate tribasic hydrate, reagentplus r, trisodium 2-hydroxypropane-1,2,3-tricarboxylate hydrate | |
HRXKRNGNAMMEHJ-UHFFFAOYSA-K | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate | |
21868242 | |
0.998 | |
Crystalline Powder or Crystals |
RUO – Research Use Only