Fisherbrand™ Clamp, Tri-Grip
Clamp designed to securely hold every type of laboratory glassware and apparatus.
$12.91 - $18.50
Specifications
Autoclavable Autoclavable
Type Mounting Clip, Reducer
Material Zinc Alloy
Color Silver
Jaw Type 3-Prong Dual Adjust
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
01-257-572
View Documents
Fisherbrand™
FW20030
Small
Each for $12.91
Case of 80 Each for $1,032.80
01-257-573
View Documents
Fisherbrand™
FW20070
Large
Each for $18.50
Case of 50 Each for $925.00
Description
Description
Designed to securely hold a large variety of glassware and apparatus.
  • Designed to securely hold a large variety of glassware and apparatus.
  • Ideal for irregularly shaped objects.
  • Utilizing thumbscrews, both prongs are easily adjusted over a wider range.
  • Dual adjustable clamps are offered in chrome plated zinc.
  • Supplied with non-slip vinyl sleeves.
Specifications
Specifications
Autoclavable
Zinc Alloy
3-Prong Dual Adjust
Mounting Clip, Reducer
Silver
Non-sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'