Fisherbrand™ Clear Boston Round Glass Bottles without Caps
Type III soda-lime glass is ideal for solvent, chemical or sample storage
 View more versions of this product
Catalog No. FB02911939
Description
Description
  • General use bottles ideal for when content integrity and sample or product visibility is important
  • Narrow screw neck opening, curved shoulders, and a round body
  • Clear glass offers minimal chemical interference with contents and is appropriate when visibility is important but protection from UV rays is not necessary
  • Environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage
Specifications
Specifications
480 mL
Clear
16 mm
60
No
168 mm
28-400 mm
16 oz.
25 mm
Glass (Type 3)
28-400
Narrow
Boston Round Bottle
75 mm