Fisherbrand™ Clear Standard Wide Mouth Glass Bottles with White Polypropylene Caps
Clear, Type III soda-lime glass ideal for general, small to large volume storage
 View more versions of this product
Catalog No. FB02911913
Explore available promotions
Description
Description
  • Features a wide mouth opening for easy transfer of contents
  • Applications include pharmaceutical, chemical, environmental, and food packaging and storing
  • Manufactured from clear flint glass
Specifications
Specifications
950 mL
Clear
12
70-400
Wide
PTFE/Polyethylene Foam
70-400 mm
32 oz.
Glass (Type 3)
White
Yes
Standard Wide Mouth Bottle
Polypropylene