missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Clindamycin, White Powder, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219116880
Specifications
Clindamycin | |
Solid | |
White | |
C18H34Cl2N2O5S | |
(7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin | |
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl | |
461.439 | |
461.4 |
Clindamycin | |
21462-39-5 | |
100 mg | |
MFCD07793327 | |
AUODDLQVRAJAJM-PIPVPTKYSA-N | |
(2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride | |
134129519 |
Chemical Identifiers
21462-39-5 | |
461.439 | |
AUODDLQVRAJAJM-PIPVPTKYSA-N | |
134129519 | |
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl |
C18H34Cl2N2O5S | |
MFCD07793327 | |
(7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin | |
(2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride |
Safety and Handling
Recommended Storage : Store at 4°C.