missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Clindamycin, White Powder, MP Biomedicals™

$710.08 - $710.08

Chemical Identifiers

CAS 21462-39-5
Molecular Formula C18H34Cl2N2O5S
Molecular Weight (g/mol) 461.439
MDL Number MFCD07793327
InChI Key AUODDLQVRAJAJM-PIPVPTKYSA-N
Synonym (7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin
PubChem CID 134129519
IUPAC Name (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19116880
View Documents
MP Biomedicals Inc
0219116880
100 mg
Each for $710.08
 
Description

Description

Specifications

Chemical Identifiers

21462-39-5
461.439
AUODDLQVRAJAJM-PIPVPTKYSA-N
134129519
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl
C18H34Cl2N2O5S
MFCD07793327
(7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin
(2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride

Specifications

Clindamycin
21462-39-5
100 mg
MFCD07793327
AUODDLQVRAJAJM-PIPVPTKYSA-N
(2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
134129519
Clindamycin
Solid
White
C18H34Cl2N2O5S
(7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl
461.439
461.4
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C.

SDS
missing translation for 'documents'

missing translation for 'documents'