missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) acetate, anhydrous, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2321830

 View more versions of this product

Catalog No. AAB2321830


Explore available promotions

Description

Description

Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

Solubility
Soluble in water, alcohol and dilute acids.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Cobalt(II) acetate
71-48-7
Odorless
Co(OOCCH3)2
250 g
Hygroscopic
cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate
QAHREYKOYSIQPH-UHFFFAOYSA-L
cobalt(2+);diacetate
6277
177.02
anhydrous
298°C (decomposition)
C4H6CoO4
MFCD00008689
UN3077
14,2433
Soluble in water,alcohol and dilute acids.
[Co++].CC([O-])=O.CC([O-])=O
177.02
CHEBI:85138
≥98%

Chemical Identifiers

71-48-7
177.02
QAHREYKOYSIQPH-UHFFFAOYSA-L
6277
cobalt(2+);diacetate
C4H6CoO4
MFCD00008689
cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate
CHEBI:85138
[Co++].CC([O-])=O.CC([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H341-H350i-H360F

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 200-755-8

RTECSNumber : AG3150000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only