Learn More
Cobalt(II) acetate, anhydrous, 98+%, Thermo Scientific Chemicals
$447.41 - $1765.92
Chemical Identifiers
CAS | 71-48-7 |
---|---|
Molecular Formula | C4H6CoO4 |
Molecular Weight (g/mol) | 177.02 |
MDL Number | MFCD00008689 |
InChI Key | QAHREYKOYSIQPH-UHFFFAOYSA-L |
Synonym | cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate |
PubChem CID | 6277 |
ChEBI | CHEBI:85138 |
IUPAC Name | cobalt(2+);diacetate |
SMILES | [Co++].CC([O-])=O.CC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2321818
|
Thermo Scientific Chemicals
B2321818 |
50 g |
Each for $447.41
|
|
|||||
AAB2321830
|
Thermo Scientific Chemicals
B2321830 |
250 g |
Each for $1,765.92
|
|
|||||
Description
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.
Solubility
Soluble in water, alcohol and dilute acids.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
71-48-7 | |
177.02 | |
QAHREYKOYSIQPH-UHFFFAOYSA-L | |
6277 | |
cobalt(2+);diacetate |
C4H6CoO4 | |
MFCD00008689 | |
cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate | |
CHEBI:85138 | |
[Co++].CC([O-])=O.CC([O-])=O |
Specifications
71-48-7 | |
Odorless | |
Co(OOCCH3)2 | |
50 g | |
Hygroscopic | |
cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate | |
QAHREYKOYSIQPH-UHFFFAOYSA-L | |
cobalt(2+);diacetate | |
6277 | |
177.02 | |
Cobalt(II) acetate, anhydrous |
298°C (decomposition) | |
C4H6CoO4 | |
MFCD00008689 | |
UN3077 | |
14,2433 | |
Soluble in water,alcohol and dilute acids. | |
[Co++].CC([O-])=O.CC([O-])=O | |
177.02 | |
CHEBI:85138 | |
≥98% |
Safety and Handling
GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H341-H350i-H360F
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 200-755-8
RTECSNumber : AG3150000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only