missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) acetate tetrahydrate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 293060010

 View more versions of this product

Catalog No. AC293060010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt(II) acetate tetrahydrate
97%
Pink to Pink-Red
1 kg
C4H6CoO4·4H2O
MFCD00149569
15, 2417
Solubility in water: 380g/L (20°C). Other solubilities: soluble in acids,soluble in alcohols,soluble in pentyl acetate
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
249.08
249.08
Plastic bottle
140.0°C
6147-53-1
Crystalline Powder and/or Crystals
98% min. (Complexometry)
(CH3CO2)2Co·4H2O
04,99; 16,95
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
cobalt(2+);diacetate;tetrahydrate
61372
≥98%

Chemical Identifiers

6147-53-1
249.08
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
61372
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
C4H6CoO4·4H2O
MFCD00149569
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
cobalt(2+);diacetate;tetrahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Suspected of causing genetic defects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer by inhalation.
May damage fertility.<br/

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Obtain speci

GHS Signal Word: Danger

RTECSNumber : AG3325000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only