Learn More
Cobalt(II) acetate tetrahydrate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 293060500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cobalt(II) acetate tetrahydrate | |
97% | |
Pink to Pink-Red | |
50 g | |
C4H6CoO4·4H2O | |
MFCD00149569 | |
15, 2417 | |
Solubility in water: 380g/L (20°C). Other solubilities: soluble in acids, soluble in alcohols, soluble in pentyl acetate | |
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2] | |
249.08 | |
249.08 | |
Plastic bottle |
140.0°C | |
6147-53-1 | |
Crystalline Powder and/or Crystals | |
98% min. (Complexometry) | |
(CH3CO2)2Co·4H2O | |
04,99; 16,95 | |
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441 | |
ZBYYWKJVSFHYJL-UHFFFAOYSA-L | |
cobalt(2+);diacetate;tetrahydrate | |
61372 | |
≥98% |
Chemical Identifiers
6147-53-1 | |
249.08 | |
ZBYYWKJVSFHYJL-UHFFFAOYSA-L | |
61372 | |
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2] |
C4H6CoO4·4H2O | |
MFCD00149569 | |
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441 | |
cobalt(2+);diacetate;tetrahydrate |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Suspected of causing genetic defects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer by inhalation.
May damage fertility.<br/
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Obtain speci
GHS Signal Word: Danger
RTECSNumber : AG3325000
RUO – Research Use Only