missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) hydroxide, 95%, pure, Thermo Scientific Chemicals

$185.61 - $660.09

Chemical Identifiers

CAS 21041-93-0
Molecular Formula CoH2O2
Molecular Weight (g/mol) 92.95
MDL Number MFCD00016015
InChI Key ASKVAEGIVYSGNY-UHFFFAOYSA-L
Synonym cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
PubChem CID 10129900
SMILES [OH-].[OH-].[Co++]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC453892500
SDS View Documents
Thermo Scientific Chemicals
453892500
250 g N/A
 
AC453890010
SDS View Documents
Thermo Scientific Chemicals
453890010
1 kg N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

21041-93-0
92.95
ASKVAEGIVYSGNY-UHFFFAOYSA-L
10129900
CoH2O2
MFCD00016015
cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
[OH-].[OH-].[Co++]

Specifications

21041-93-0
95%
Co(OH)2
15,2426
ASKVAEGIVYSGNY-UHFFFAOYSA-L
λ²-cobalt(2+) dihydroxide
10129900
95%
Cobalt(II) hydroxide
250 g
CoH2O2
MFCD00016015
cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
[OH-].[OH-].[Co++]
92.95
92.95
Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Very toxic to aq

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:

GHS Signal Word: Danger

EINECSNumber : 244-166-4

SDS
Documents

Documents

RUO – Research Use Only