missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Coenzyme A Trilithium Salt Dihydrate ≥96%, White Crystalline Powder, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210049350
Specifications
Coenzyme A Trilithium Salt Dihydrate >96% | |
White | |
C21H33Li3N7O16P3S | |
50 mg | |
QSCBPHBAFBVXRK-UHFFFAOYSA-K | |
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate | |
53442190 | |
Crystalline Powder |
18439-24-2 | |
≥96% | |
MFCD00075848 | |
coenzyme a, trilithium salt | |
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O | |
785.329 | |
821.4 |
Chemical Identifiers
18439-24-2 | |
785.329 | |
QSCBPHBAFBVXRK-UHFFFAOYSA-K | |
53442190 | |
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O |
C21H33Li3N7O16P3S | |
MFCD00075848 | |
coenzyme a, trilithium salt | |
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate |
Safety and Handling
Recommended Storage : Store at -20 °C.