missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Coenzyme A Trilithium Salt Dihydrate ≥96%, White Crystalline Powder, MP Biomedicals™

$114.01 - $2523.25

Chemical Identifiers

CAS 18439-24-2
Molecular Formula C21H33Li3N7O16P3S
Molecular Weight (g/mol) 785.329
MDL Number MFCD00075848
InChI Key QSCBPHBAFBVXRK-UHFFFAOYSA-K
Synonym coenzyme a, trilithium salt
PubChem CID 53442190
IUPAC Name trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
SMILES [Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10049310
View Documents
MP Biomedicals Inc
0210049310
10 mg
Each for $114.01
 
ICN10049325
View Documents
MP Biomedicals Inc
0210049325
25 mg
Each for $193.93
 
ICN10049350
View Documents
MP Biomedicals Inc
0210049350
50 mg
Each for $308.35
 
ICN10049380
View Documents
MP Biomedicals Inc
0210049380
100 mg
Each for $444.51
 
ICN10049383
View Documents
MP Biomedicals Inc
0210049383
250 mg
Each for $891.60
 
ICN10049390
View Documents
MP Biomedicals Inc
0210049390
500 mg
Each for $2,523.25
 
Description

Description

Specifications

Chemical Identifiers

18439-24-2
785.329
QSCBPHBAFBVXRK-UHFFFAOYSA-K
53442190
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O
C21H33Li3N7O16P3S
MFCD00075848
coenzyme a, trilithium salt
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate

Specifications

18439-24-2
≥96%
MFCD00075848
coenzyme a, trilithium salt
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O
785.329
821.4
Coenzyme A Trilithium Salt Dihydrate >96%
White
C21H33Li3N7O16P3S
10 mg
QSCBPHBAFBVXRK-UHFFFAOYSA-K
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
53442190
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20 °C.

SDS
missing translation for 'documents'

missing translation for 'documents'