Conductivity Solution, Oakton™
Use this conductivity solution for accurate test results in the field or lab. Oakton™ Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
Supplier: Oakton Instruments WD0065323
Description
Use this conductivity solution for accurate test results in the field or lab. OaktonTM Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
- Features temperature corrections printed on the bottle
- Certified values listed in the tables and are referenced to 25°C
- Two-year shelf-life on all solutions
- Conductivity-to-TDS Calibration Values: ppm NaCl: 10.7, ppm 442: 14.74, ppm KCl: 11.6
Specifications
Cole-Parmer Conductivity Standards | |
Conductivity Standard | |
Conductivity Standard | |
0.023 mS (23 μS) |