Conductivity Solution, Oakton™
Use this conductivity solution for accurate test results in the field or lab. Oakton™ Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
$59.35 - $65.37
Specifications
Chemical Name or Material Cole-Parmer Conductivity Standards
Type Conductivity Standard
View More Specs
Products 5
Catalog Number Mfr. No. Conductivity Price Quantity  
Catalog Number Mfr. No. Conductivity Price Quantity  
13-300-114
View Documents
Oakton Instruments
WD0065323
0.023 mS (23 μS)
Each for $65.37
13-300-111
View Documents
Oakton Instruments
0065316
0.084 mS (84 μS)
Each for $62.44
13-300-117
View Documents
Oakton Instruments
65347
0.447 mS (447 μS)
Each for $59.35
13-300-112
View Documents
Oakton Instruments
ZZ0065318
1.413 mS (1413 μS)
Each for $62.26
13-300-110
View Documents
Oakton Instruments
0065315
1.5 mS (1500 μS)
Each for $61.56
Description
Description
Use this conductivity solution for accurate test results in the field or lab. OaktonTM Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
  • Features temperature corrections printed on the bottle
  • Certified values listed in the tables and are referenced to 25°C
  • Two-year shelf-life on all solutions
  • Certificates of analysis available separately
Specifications
Specifications
Cole-Parmer Conductivity Standards
Conductivity Standard
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'