Conductivity Solution, Oakton™
Use this conductivity solution for accurate test results in the field or lab. Oakton™ Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
Supplier:  Oakton Instruments 65347
 View more versions of this product
Catalog No. 13-300-117
Explore available promotions
Description
Description
Use this conductivity solution for accurate test results in the field or lab. OaktonTM Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
  • Features temperature corrections printed on the bottle
  • Certified values listed in the tables and are referenced to 25°C
  • Two-year shelf-life on all solutions
  • Conductivity-to-TDS Calibration Values: ppm NaCl: 215.5, ppm 442: 300, ppm KCl: 225.6
Specifications
Specifications
Cole-Parmer Conductivity Standards
Conductivity Standard
Conductivity Standard
0.447 mS (447 μS)