Fisherbrand™ Complete Plumbing Assemblies for Lab Workstations
Fisherbrand™ Complete Plumbing Assemblies are designed to be used with Lab Workstations.
Description
- Service turret is color coded based on service
- Service turret is a 135 degree turret with one polished chrome valve with a brass floating cone and integral brass seat.
- Service turrets provide metering control of laboratory gases at pressures up to 60 PSI.
- System service hoses carry services from teh overhead service panel through the upright of the table and to the shank of the fixture. Hoses extend 48 in. above the workstation.
- System service hoses are color coded reinforced PVC
- Male Quick Connects has a double end shut off plug that have an internal valve that shuts off when uncoupled.
Specifications
Left Side, Nitrogen Service Complete Plumbing Assembly | |
PVC, Chrome | |
Male Quick Connect, System Hose and Service Turret |
Brown | |
Nitrogen Service Plumbing Assembly | |
Left side |