Contec™ Polynit™ Heatseal Wipes
Use these wipes for all ultra-critical applications and general purpose cleaning. Contec™ Polynit Heatseal Wipes are constructed of 100% textured knitted polyester. Wipe are laser-cut, which seals the edges of the fabric for complete utilization of the wipe’s surface.
$87.09 - $1301.45
Specifications
Material Knitted Polyester
Autoclavable Not Autoclavable
Disposable Single-use
View More Specs
Products 9
Catalog Number Mfr. No. Product Type Sterility Dimensions (L x W) Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Product Type Sterility Dimensions (L x W) Packaging Quantity Price Quantity  
19-038-562
View Documents
Contec™
PNHS44
Heatseal Wipe Non-sterile 10 x 10 cm (4 x 4 in.) 300 Wipes/Bag (Stack Packed)
Pack of 300 Each for $95.35
Case of 16 Pack for $1,525.60
19038600
View Documents
Contec™
PNHS44B
Heatseal Wipe Non-sterile 10 x 10 cm (4 x 4 in.) 600 Wipes/Bag (Bulk)
Case of 7200 Each for $1,301.45
19-170-943
View Documents
Contec™
LWPS0006
Heatseal Wipe Sterile 23 x 23 cm (9 x 9 in.) 25 Wipes/Inner Bag, 4 Inner Bags/Bag
Pack of 100 Each for $1,301.45
Case of 12 Pack for $2,672.16
29-636-4382
View Documents
Contec™
PNHS99
Heatseal Wipe Non-sterile 23 x 23 cm (9 x 9 in.) 150 Wipes/Bag (Stack Packed)
Case of 150 Each for $854.68
19047513
View Documents
Contec™
PNHS99B150
Heatseal Wipe Non-sterile 23 x 23 cm (9 x 9 in.) 150 Wipes/Bag (Bulk)
Pack of 150 Each for $87.09
Case of 8 Pack for $696.72
19-033-768
View Documents
Contec™
PNHS99B
Heatseal Wipe Non-sterile 23 x 23 cm (9 x 9 in.) 300 Wipes/Bag (Bulk)
Pack of 300 Each for $185.94
Case of 4 Pack for $743.76
19-033741B
View Documents
Contec™
PNHS1212
Heatseal Wipe Non-sterile 30 x 30 cm (12 x 12 in.) 75 Wipes/Bag (Stack Packed)
Pack of 75 Each for $90.79
Case of 10 Pack for $907.90
19-170-944
View Documents
Contec™
PNHS1212B
Heatseal Wipe Non-sterile 30 x 30 cm (12 x 12 in.) 75 Wipes/Bag (Bulk)
Case of 750 Each for $769.38
19-161-961
View Documents
Contec™
LWPS0007
Heatseal Wipe Sterile 30 x 30 cm (12 x 12 in.) 1000 Wipes/Bag (Stacked)
Case of 1000 Each for $769.38
Description
Description
  • Textured 100% polyester no-run knit wipes with heat-sealed edges
  • Laser-cutting seals fibers at wipe's edge and allows for 100% utilization of wipe
  • Recommended for ISO Class 3-8 environments
  • Extremely low in particles, fibers and non-volatile residues (NVRS)
  • Good sorbency with solvents
  • Abrasion and chemical resistant
  • Laundered and packaged in an ISO Class 4 cleanroom
  • Available Validated Sterile to a 10-6 Sterility Assurance Level in accordance with standards and recommended practices of the AAMI/ISO 11137:2006 Guidelines
Order Info
Available in custom sizes and shapes. Contact your Fisher Safety Customer Service Representative for details.
Specifications
Specifications
Knitted Polyester
Not Autoclavable
Single-use
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'