missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Contec™ Sterile 70% Isopropanol

Contec™ Sterile 70% Isopropanol is ideal for wiping down of isolators, cabinets, laminar flow hoods and surfaces.

$352.64 - $583.53

Specifications

Form Liquid
Sterility Sterile
Appearance Clear
Certifications/Compliance USP 797
Composition 70% USP Isopropyl Alcohol and 30% USP Purified Water
View More Specs

Products 3
Catalog Number Mfr. No. Volume (English) Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (English) Volume (Metric) Price Quantity  
19-033-766
View Documents
Contec™
SB167030IR
16 oz. 0.47 L
Case of 12 Each for $583.53
 
19-033-765
View Documents
Contec™
SB327030IR
32 oz. 0.94 L
Case of 12 Each for $391.97
 
19-033-775
Accessories View Documents
Contec™
SB1287030IR
1 gal. 3.8 L
Case of 4 Each for $352.64
 
Description

Description

Contains 70% by volume USP-grade Isopropanol (IPA) and 30% USP-purified water.

 

  • Evaporates and leaves extremely low residues
  • Eliminates mixing, filtration, processing and QC expense of in-house production of sterile isopropanol water blends
  • Filtered through 0.2μm in-line filter which eliminates the need for in-house filtration
  • Gamma-irradiated to a sterility assurance level (SAL) of 10-6
  • Prefiltered, packaged and sterilized for easy introduction into critical environments
  • Certificate of processing and sterility with each case
  • Induction-sealed bottles reduce opportunity for leakage during sterilization and shipment
  • 16 and 32 oz. sizes include a flexible trigger sprayer for instant adjustment for a variety of applications
  • Recyclable plastic bottle

 

Compliance

Compliant with USP 797; validated per the Association for the Advancement of Medical Instrumentation (AAMI) 11137 Guidelines to a 10-6 Sterility Assurance Level (SAL)

Specifications

Specifications

Liquid
Clear
70% USP Isopropyl Alcohol and 30% USP Purified Water
Sterile
USP 797
Wipe down of isolators, cabinets, laminar flow hoods and surfaces
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'