CoorsTek™ Porcelain Capsule-Form Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
Supplier:  CoorsTek™ CRS66048
 View more versions of this product
Catalog No. 08-108A
Explore available promotions
Description
Description
  • Suitable for ash determinations, organic combustions, ignitions
  • Capsule form with straight sides, flat bottom
  • Glazed inside and out except on rim
Specifications
Specifications
Porcelain
12 mL