Copan Diagnostics Calibrated Disposable Inoculating Loops and Needles in Zip Lock Pack
Calibrated 1 and 10μL plastic disposable inoculating loops and needles
Supplier: Copan Diagnostics COPS1
Description
Calibrated 1 and 10μL plastic disposable inoculating loops and needles
Zip-lock closure reseals the pack between use to avoid contamination. Re-sealable bag allows the user to open and close the loop pack to remove individual loops.
- Disposable, sterile plastic loops and needles manufactured from soft-flexible or hard-rigid plastic
- Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
- Ultrasmooth surfaces allow easy streaking without cutting or gouging agar surface
- Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
- Color coded for easy product identification
- Packed in tamper-proof bags to guarantee sterility
- Certificate of calibration included in each case
Specifications
Single-use | |
Plastic | |
Light Green | |
Evans blue dye certificate of calibration | |
Yes |
Sterile | |
1 μL | |
Inoculating Loop, Flexible | |
Tamperproof Ziplock resealable bag |