Copan Diagnostics Calibrated Disposable Inoculating Loops and Needles in Zip Lock Pack
Calibrated 1 and 10μL plastic disposable inoculating loops and needles
$636.62 - $642.13
Specifications
Material Plastic
Sterility Sterile
Disposable Single-use
Includes Evans blue dye certificate of calibration
Packaging Type Tamperproof Ziplock resealable bag
View More Specs
Products 3
Catalog Number Mfr. No. Color Product Type Price Quantity  
Catalog Number Mfr. No. Color Product Type Price Quantity  
OXCOPS1
View Documents
Copan Diagnostics
COPS1
Light Green Inoculating Loop, Flexible
Pack of 1000 Each for $636.62
OXCOPS10
View Documents
Copan Diagnostics
NCO0014
Light Blue Inoculating Loop, Flexible
Pack of 1000 Each for $642.13
OXCOPH10
View Documents
Copan Diagnostics
COPH10
Dark Blue Inoculating Loop, Rigid
Pack of 1000 Each for $642.13
Description
Description
Calibrated 1 and 10μL plastic disposable inoculating loops and needles
Zip-lock closure reseals the pack between use to avoid contamination. Re-sealable bag allows the user to open and close the loop pack to remove individual loops.
  • Disposable, sterile plastic loops and needles manufactured from soft-flexible or hard-rigid plastic
  • Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
  • Ultrasmooth surfaces allow easy streaking without cutting or gouging agar surface
  • Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
  • Color coded for easy product identification
  • Packed in tamper-proof bags to guarantee sterility
  • Certificate of calibration included in each case
Specifications
Specifications
Plastic
Single-use
Tamperproof Ziplock resealable bag
Sterile
Evans blue dye certificate of calibration
Yes
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'