Copan Diagnostics Calibrated Disposable Inoculating Loops and Needles in Zip Lock Pack
Calibrated 1 and 10μL plastic disposable inoculating loops and needles
Supplier:  Copan Diagnostics NCO0014
 View more versions of this product
Catalog No. OXCOPS10
Explore available promotions
Description
Description
Calibrated 1 and 10μL plastic disposable inoculating loops and needles
Zip-lock closure reseals the pack between use to avoid contamination. Re-sealable bag allows the user to open and close the loop pack to remove individual loops.
  • Disposable, sterile plastic loops and needles manufactured from soft-flexible or hard-rigid plastic
  • Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
  • Ultrasmooth surfaces allow easy streaking without cutting or gouging agar surface
  • Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
  • Color coded for easy product identification
  • Packed in tamper-proof bags to guarantee sterility
  • Certificate of calibration included in each case
Specifications
Specifications
Single-use
Plastic
Light Blue
Evans blue dye certificate of calibration
Yes
Sterile
10 μL
Inoculating Loop, Flexible
Tamperproof Ziplock resealable bag