missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(I) bromide-dimethyl sulfide complex, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 213180250

 View more versions of this product

Catalog No. AC213180250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper(I) bromide-dimethyl sulfide complex
54678-23-8
Beige-Gray to White or Green
98.5% min. (Iodometry)
C2H6BrCuS
MFCD00043295
copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i
[Cu]Br.CSC
205.58
205.58
Crystalline Powder and/or Chunks
99%
132°C
Authentic
Glass bottle
(CH3)2S·CuBr
25 g
PMHQVHHXPFUNSP-UHFFFAOYSA-M
(methylsulfanyl)methane; bromocopper
2724234
99%

Chemical Identifiers

54678-23-8
205.58
PMHQVHHXPFUNSP-UHFFFAOYSA-M
2724234
C2H6BrCuS
MFCD00043295
copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i
[Cu]Br.CSC
Videos
Safety and Handling

Safety and Handling

AIR SENSITIVE

MOISTURE SENSITIVE

EINECSNumber : 259-292-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only