missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(I) iodide, 99.995%, (trace metal basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 201500050

 View more versions of this product

Catalog No. AC201500050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, for ultra trace analysis
Specifications

Specifications

1290°C
605°C
7681-65-4
Powder
31.0 to 34% (Cu) (Complexometry)
CuI
01,169; 02,92; 03,69; 05,167; 06,147; 07,81; 08,121; 09,124; 10,107; 11,141; 12,141; 16,98; 17,88
cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g
190.45
100.00%
Glass bottle
Copper(I) iodide
99.995%
Brown or Gray to White
5 g
CuI
MFCD00010978
15, 2652
Solubility in water: insoluble. Other solubilities: soluble in ammonia and potassium iodide solutions,practically insoluble in alcohol
190.45
Trace Metal Basis
5.62

Chemical Identifiers

7681-65-4
190.45
cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g
CuI
MFCD00010978
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut

GHS Signal Word: Danger

EINECSNumber : 231-674-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only