Learn More
Copper(I) iodide, 99.995%, (trace metal basis), Thermo Scientific Chemicals
$83.36 - $83.36
Chemical Identifiers
CAS | 7681-65-4 |
---|---|
Molecular Formula | CuI |
Molecular Weight (g/mol) | 190.45 |
MDL Number | MFCD00010978 |
Synonym | cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC201500050
|
Thermo Scientific Chemicals
201500050 |
5 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, for ultra trace analysisChemical Identifiers
7681-65-4 | |
190.45 | |
cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g |
CuI | |
MFCD00010978 |
Specifications
1290°C | |
7681-65-4 | |
Powder | |
31.0 to 34% (Cu) (Complexometry) | |
CuI | |
01,169; 02,92; 03,69; 05,167; 06,147; 07,81; 08,121; 09,124; 10,107; 11,141; 12,141; 16,98; 17,88 | |
cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g | |
190.45 | |
100.00% | |
Glass bottle | |
Copper(I) iodide, 99.995% |
605°C | |
Brown or Gray to White | |
5 g | |
CuI | |
MFCD00010978 | |
15, 2652 | |
Solubility in water: insoluble. Other solubilities: soluble in ammonia and potassium iodide solutions,practically insoluble in alcohol | |
190.45 | |
Trace Metal Basis | |
5.62 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
GHS Signal Word: Danger
EINECSNumber : 231-674-6
RUO – Research Use Only