missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) acetate monohydrate, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 423591000

 View more versions of this product

Catalog No. AC423591000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

240.0°C
115.0°C
Blue-Green or Green
100 g
(CH3CO2)2Cu·H2O
01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 14,199; 15,99; 17,176
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper;diacetate;hydrate
165397
98.0 to 102.0%
Authentic
1.88g/mL
Copper(II) acetate monohydrate
6046-93-1
Solid
C4H8CuO5
MFCD00149570
15,2614
Solubility in water: 72g/L (20°C). Other solubilities: soluble in alcohol and ether
O.[Cu++].CC([O-])=O.CC([O-])=O
199.65
199.65
ACS Reagent
Plastic bottle

Chemical Identifiers

6046-93-1
199.65
NWFNSTOSIVLCJA-UHFFFAOYSA-L
165397
O.[Cu++].CC([O-])=O.CC([O-])=O
C4H8CuO5
MFCD00149570
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
copper;diacetate;hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only