missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) acetate monohydrate, ACS reagent, Thermo Scientific Chemicals

$97.40 - $195.20

Chemical Identifiers

CAS 6046-93-1
Molecular Formula C4H8CuO5
Molecular Weight (g/mol) 199.65
MDL Number MFCD00149570
InChI Key NWFNSTOSIVLCJA-UHFFFAOYSA-L
Synonym copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
PubChem CID 165397
IUPAC Name copper;diacetate;hydrate
SMILES O.[Cu++].CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC423591000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
423591000
100 g Plastic bottle
Each for $97.40
 
AC423595000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
423595000
500 g Plastic bottle
Each for $195.20
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

6046-93-1
199.65
NWFNSTOSIVLCJA-UHFFFAOYSA-L
165397
O.[Cu++].CC([O-])=O.CC([O-])=O
C4H8CuO5
MFCD00149570
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
copper;diacetate;hydrate

Specifications

240.0°C
6046-93-1
Solid
C4H8CuO5
MFCD00149570
15,2614
Solubility in water: 72g/L (20°C). Other solubilities: soluble in alcohol and ether
O.[Cu++].CC([O-])=O.CC([O-])=O
199.65
199.65
ACS Reagent
Plastic bottle
Copper(II) acetate monohydrate
115.0°C
Blue-Green or Green
100 g
(CH3CO2)2Cu·H2O
01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 14,199; 15,99; 17,176
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper;diacetate;hydrate
165397
98.0 to 102.0%
Authentic
1.88g/mL
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only