Learn More
Copper(II) bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals
$79.03 - $431.00
Chemical Identifiers
CAS | 7789-45-9 |
---|---|
Molecular Formula | Br2Cu |
Molecular Weight (g/mol) | 223.36 |
MDL Number | MFCD00010970 |
Synonym | Cupric bromide |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC206330050
|
Thermo Scientific Chemicals
206330050 |
5 g | Glass bottle |
Each for $79.03
|
|
||||
AC206331000
|
Thermo Scientific Chemicals
206331000 |
100 g | Glass bottle |
Each for $120.13
|
|
||||
AC206335000
|
Thermo Scientific Chemicals
206335000 |
500 g | Glass bottle |
Each for $431.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
7789-45-9 | |
223.36 | |
Cupric bromide |
Br2Cu | |
MFCD00010970 |
Specifications
900.0°C | |
7789-45-9 | |
Crystalline Powder | |
99% min. (Iodometry) | |
CuBr2 | |
01,161; 02,84; 05,158; 06,138; 13,112; 15,100 | |
Cupric bromide | |
223.36 | |
≥99% | |
Glass bottle | |
Copper(II) bromide |
498.0°C | |
Black-Gray to Gray | |
5 g | |
Br2Cu | |
MFCD00010970 | |
15, 2619 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, acetone, nh3, practically insoluble in benzene, diethyl ether,, conc. h2so4 | |
223.36 | |
Extra Pure | |
4.77 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 232-167-2
TSCA : TSCA
RUO – Research Use Only